Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Universal Plus Pads
Color-coded chemical absorbents for safe clean-up of hazardous fluids
$302.00 - $731.50
Specifications
Certifications/Compliance | RoHS |
---|---|
Color | Green |
Material | Polypropylene |
Type | Universal Plus Pad |
For Use With (Application) | Aggressive chemicals and hazardous fluids |
Catalog Number | Mfr. No. | Dimensions (L x W) | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Packaging Quantity | Price | Quantity & Availability | ||||
19-150-258
|
Brady™
SPCUN1719 |
19 x 17 in. (48.2 x 43 cm) | 100/Pk. |
Case of 100 for $731.50
|
|
||||
19-150-257
|
Brady™
SPCUN1113 /DEL |
12 x 12 in. (30.48 x 30.48 cm) | 200/Pk. |
Case of 200 for $302.00
|
|
||||
19-041-053
|
Brady™
SPCUN111350 |
12 x 12 in. (30.48 x 30.48 cm) | 50/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Chemically inert and ideal for aggressive fluids
- Intended for use with chemicals and hazardous materials
- Universal meltblown polypropylene
- Three-ply construction makes it stronger, more durable and reduces lint
- Dimpled
- Medium weight
SPCUN1719 is perforated.
Specifications
RoHS | |
Polypropylene | |
Aggressive chemicals and hazardous fluids | |
Medium-weight |
Green | |
Universal Plus Pad | |
3-ply |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title