Brady™ Universal Plus Pads Available on GSA/VA Contract for Federal Government customers only.

Color-coded chemical absorbents for safe clean-up of hazardous fluids

Manufacturer: Brady™ SPCUN1113

 View more versions of this product

Catalog No. 19-150-257

  Check Availability
Add to Cart

    Specifications

    Product Type Universal Plus Pad
    Capacity (English) 20 gal.
    Capacity (Metric) 75.7L
    Length (Metric) 30.48cm
    Length (English) 12 in.
    For Use With (Application) For use with aggressive chemicals and hazardous fluids
    Material Polypropylene
    Weight Capacity Medium
    Thickness 3 ply
    Width (English) 12 in.
    Width (Metric) 30.48cm
    Capacity 20 gal., 75.7L
    Dimensions (L x W) 12 x 12 in. (30.48 x 30.48cm)
    Quantity 200/Pk
    Absorbancy Medium
    Certifications/Compliance RoHS
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit