Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Universal Plus Rolls

Universal Plus Rolls are chemically inert and green in color (the universal symbol for safety) to indicate that they are intended for use with hazardous materials.

$284.50

Specifications

Color Green
Product Type Dimpled Absorbent Roll
For Use With (Application) Aggressive fluids, overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
Includes Universal symbol for safety
Length (English) 150 ft.
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (English) Quantity Weight (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Quantity Weight (English) Width (English) Price Quantity & Availability  
19-150-262
View Documents
Brady™
SPCUN3800
40 gal. Each 27 lb. 30 in. Case for $284.50
Add to cart
 
Description

Description

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Medium weight - optimal for most general maintenance needs
  • Universal Plus products feature 3-ply construction increasing durability while reducing lint
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications

Specifications

Green
Aggressive fluids, overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
150 ft.
Dimpled Absorbent Roll
Universal symbol for safety
Meltblown Polypropylene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit