Learn More
Brady™ UXT Xtra Tough Universal Heavy Rolls
Made to handle virtually any liquid (water, petroleum and chemical-based)
Manufacturer:  Brady™ UXT30DP
 View more versions of this product
Catalog No. 17-111-203
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Three-ply construction
  • Double-sided, low-linting coverstock and meltblown core
  • Abrasion resistant and non-abrasive performance
  • Perforated
Description & Specifications
Specifications
Heavy
Food Processing, High Tech Industries, Machining of small parts, Paint room, Pharmaceutical, Printing
1/Cs.
UXT Xtra Tough Universal Heavy Roll
45.7m x 76.2cm (150 ft. x 30 in.)
35 gal./bale