Learn More
Brady™ Vinyl Tapes (B-595) for BMP71 Printer - Clear
Indoor/Outdoor vinyl grade conforms to irregular, smooth or rough surfaces
Manufacturer:  Brady™ M71C2000595CL
 View more versions of this product
Catalog No. 17-111-033
  Check Availability
Add to cart
Specifications
Description
  • Vinyl film with permanent adhesive
  • Adheres well to pipes, walls, equipment, windows - most clean, dry surfaces
  • Resistant to grease, oil and most industrial chemicals
  • Five year outdoor durability in temperatures from 82° to -40°C (180° to -40°F, can be applied as low as -18°C (0°F)
Description & Specifications
Specifications
Clear
Labeling pipes, walls, equipment, windows
50 ft.
2 in.
Vinyl (B-595)
1/ea
15.24m x 5.08cm (50 ft. x 2 in.)
BMP71 Printer
15.24m
5.08cm
Vinyl Tape