Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ “Warning: Authorized Personnel Only” Signs
Identify areas designated for certain personnel and direct facility traffic
Supplier: Brady™ 22194
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Available in plastic, self sticking polyester and premium fiberglass materials
- Black on orange
- Language: english
- Available sizes (W x H): 10 x 7 in. or 14 x 10 in.
Specifications
ANSI Z535, OSHA 1910.145, ASTM D 1593 | |
1 Ea. | |
Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs and equipment warning signs, recommended for both indoor and outdoor use | |
25.4 cm | |
-40°F | |
14 in. | |
-40°C to 80°C | |
Warning Authorized Personnel Only | |
Black on Orange |
Plastic | |
Surface Mount | |
10 in. | |
175°F | |
0.06 in. | |
35.5 cm | |
-40°F to 175°F | |
Admittance Sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction