Brady™ “Warning: Lead Work Area -- POISON...” Signs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Brady™ 25740

 View more versions of this product

Catalog No. 19-102299

  Check Availability
Add to Cart

    Specifications

    Material Plastic
    Temperature Range (English) -40 to 175°F,-40 to 175°F
    Width (English) 10 in.,10 in.
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 7 in.,7 in.
    Certifications/Compliance ANSI Z535, OSHA 1910.145
    Message Warning Lead Work Area Poison No Smoking or Eating
    Quantity Each
    Style Surface Mount
    Temperature Range (Metric) -40 to 80°C,-40 to 80°C
    Thickness 0.06 in.,0.06 in.
    Color Black on Orange,Black on Orange
    Type Chemical and hazardous materials sign

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit