Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ “Warning: Lead Work Area -- POISON...” Signs
“Warning: Lead Work Area -- POISON...” safety signs from Brady™ help to manage and sustain a clean and safe workplace and are available in a variety of materials and sizes that best fit your application.
$31.50 - $94.00
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
---|---|
Quantity | 1 Ea. |
Adhesive Type | Non-adhesive |
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
19-036-199
|
Brady™
76085 |
Fiberglass | 10 x 14 in. |
Each for $94.00
|
|
||||
19-102299
|
Brady™
25740 |
Plastic | 7 x 10 in. |
Each for $31.50
|
|
||||
Description
- Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
- Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
Specifications
ANSI Z535, OSHA 1910.145 | |
Non-adhesive | |
-40°F | |
Chemical and Hazardous Materials Sign |
1 Ea. | |
Surface Mount | |
Warning Lead Work Area Poison No Smoking or Eating | |
Black on Orange |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title