Learn More
Brady™ BradyGlo™ Egress Marking Tape Roll, Solid Color, Green Glow
Use Brady “Glow-in-the-Dark” Tapes for visible marking at night or in low light conditions
Manufacturer:  Brady™ 76433
Catalog No. 19-103-778
Add to cart
Description
Description
Photoluminescent Polyester
Specifications
Specifications
No Text
Safety
Phosphorescent Polyester
5 yd.
3 in.
Acrylic
Each
Glow
Marking Tape
0.008 in.
97.79 mm