Brady™ BradyGlo™ High Intensity Anti-Slip Tapes

For marking steps and landings, perimeter demarcation lines, door frames, and handrails

Manufacturer: Brady™ 114636

 View more versions of this product

Catalog No. 19-103-679

  Check Availability
Add to Cart

    Specifications

    Quantity 1/roll
    Length (English) 50 ft.
    Length (Metric) 15.2m
    Width (English) 1 in.
    Width (Metric) 2.5cm
    Adhesive B-982
    Certification Compliance IBC/IFC compliant
    Material Self sticking polyester
    Product Type High intensity anti-slip tape

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit