Learn More
Brady™ BradyGlo™ NYC Egress Marking Tape L-Shaped Corner Marks - Glow-In-The-Dark Self Sticking Polyester, Solid Color, Black on Glow
High-intensity, glow-in-the-dark material complies with all New York City indoor-use requirements for egress marking.
Manufacturer:  Brady™ 81711
Catalog No. 19-103-798
Add to cart
Description
Description
  • Non-toxic, non-radioactive, explosion-safe material features an aggressive pressure-sensitive synthetic rubber adhesive
  • Complies with RS 6-1 and 6-1A of New York Local Law 26 including photoluminescence, washability, flammability, toxic gas generation and radioactivity
  • Ideal for marking the leading edges of steps and landings, perimeter demarcation lines, door frames, and handrails
  • Engineered to provide a bright, long-lasting afterglow that helps guide people to safety
Specifications
Specifications
Safety
UL 1994;UL Listed for US
Black on Glow
Marking Tape
0.0096 in.
Roll of 264 Each
No Text
Phosphorescent Polyester
1 in.
9 in.