Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Alfa Aesar™ Bradykinin (1-5)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J63233MB
 View more versions of this product
Catalog No. AAJ63233MB
Order in Bulk
Request bulk or custom formats
Specifications
Specifications
572.67
25mg
Powder
RUO
Bradykinin (1-5)
HPLC
Biologically active fragment of bradykinin
H-Arg-Pro-Pro-Gly-Phe-OH
-30° to -10°C