Learn More
Enzo Life SciencesSupplier Diversity Partner Bradykinin receptor B2 polyclonal antibody (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI905747100
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat bradykinin receptor B2. UniProt: P25023. Species Reactivity: Mouse, Rat. Crossreactivity: Predicted species reactivity based on epitope sequence homology: guinea pig, dog. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-658-505