Learn More
Bradykinin RB2/BDKRB2 Mouse anti-Human, Alexa Fluor™ 700, Clone: 471916, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB47251N100UG
Catalog No. FB47251N100
Add to cart
This item is not returnable. View return policy
Description
Description
Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Bradykinin RB2/BDKRB2
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088
NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession # P30411
100 μg
624
Human
Supernatant
Flow Cytometry
471916
Flow Cytometry 0.25-1 μg/106 cells
P30411
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human Bradykinin RB2/BDKRB2 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b