Learn More
Bradykinin RB2/BDKRB2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 471902, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9434S100UG
Catalog No. FAB9434S100
Add to cart
This item is not returnable. View return policy
Description
Description
Bradykinin RB2/BDKRB2 Monoclonal antibody specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Bradykinin RB2/BDKRB2
471902
Flow Cytometry 0.25-1 ug/10^6 cells
P30411
B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088
NS0 mouse myeloma cell line transfected with human RB2/BDKRB2, Met1-Gln391, Accession No. P30411
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human RB2/BDKRB2 in direct ELISAs and Western blots.
Flow Cytometry
Alexa Fluor™ 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
624.0
Human