Learn More
Brady™ BradyLam™ Fiberglass with Polyester Laminate Sign: NON-POTABLE WATER
Improve workplace safety with proper signage and markings. This Brady™ laminated fiberglass sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 17583
Catalog No. 19-106-774
Add to cart
Description
Description
  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Safety
Fiberglass, Polyester
12 in. x 9 in. x 0.1102 in.
Black, Red on White
Rectangle
NON-POTABLE WATER