Learn More
Brady™ Bradylink Warning Stanchion - Solid Color - Large
Bradylink Warning Posts are made of colorful polystyrene and polyethylene and are a decorative, rust-free alternative to metal posts. Each post includes base, top and c-hook.
Supplier: Brady™ 92117
Description
- Posts are non-conductive so they can be safely used in electrically hazardous areas
- Hollow base can be filled with sand for more stable application
- Bradylink Chains sold separately
- Solid color stanchion
Specifications
White | |
16 in. | |
1 Ea. | |
Polyethylene, Polystyrene |
3 x 41 in. | |
41 in. | |
411.48 mm | |
Stanchion |