Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRAF35 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA146226

Catalog No. PIPA146226


Only null left
Add to Cart

Description

Description

The target sequence has 80% sequence homology with mouse protein. Suggested positive control: antigen standard for HMG20B (transient overexpression lysate).

Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BRAF35
Polyclonal
Unconjugated
BRAF
AW610687; BRAF25; BRAF35; BRCA2-associated factor 35; fc85b02; FLJ26127; high mobility group 20 B; high mobility group 20B; high mobility group box domain containing; high mobility group protein 20 B; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; HMG20B; Hmgx2; HMGXB2; hypothetical protein LOC553572; PP7706; pp8857; si:dkey-110c1.6; SMARCE1R; SMARCE1-related protein; SOXL; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; LOW QUALITY PROTEIN: SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; Unknown (protein for MGC:134468); wu:fc85b02; zgc:110001
Rabbit
Antigen Affinity Chromatography
RUO
10362, 673
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
P15056, Q9P0W2
BRAF, HMG20B
Residues 31-50 of the human structural DNA-binding protein BRAF35.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.