BrandTech™ Accessories for seripettor™ Bottle Top Dispensers: Filling Tubes
Manufacturer: BrandTech™ 704206
 View more versions of this product
Catalog No. 13-879-81
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Filling Tube
    For Use With (Equipment) Seripettor™ bottletop dispenser II
    Material Polypropylene
    Size (English) 9.84 in.
    Size (Metric) 250 x 6.9mm
  • Description & Specifications
Related Products
Product Content Feedback