Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ accu-jet™ pro Pipet Controller Accessories

Elevates pipet control to the next level.accu-jet™ pro Pipet Controller Accessories available for order include sterile and non-sterile replacement membrane filters, silicone adapter with PTFE check valve, NiMH Battery, 240V, 60Hz Recharger, and Wall Holder.

Manufacturer:  BrandTech™ BRAND™ 26540

 View more versions of this product

Catalog No. 04-977-156


Add to cart

Description

Description

BrandTech™ accu-jet™ pro Pipet Controller Accessories available for order include:
  • Sterile 0.2μm replacement membrane filters
  • Non-Sterile 0.2μm replacement membrane filters
  • Silicone adapter with PTFE check valve
  • Recharger, 240V, 60Hz
  • NiMH Battery for cordless, untethered use.
  • Wall Holder saves space.
Specifications

Specifications

Wall Support
accu-jet™ pro Pipet Controller
Gray
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit