Learn More
BrandTech™ Analog Dispensette™ Organic Bottle-Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
Ideal for HPLC and combinatorial chemistry labs
Manufacturer:  BrandTech™ 4731141
 View more versions of this product
Catalog No. 13-688-272
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
1 to 10mL
Yes
≤0.1% 10μL
Up to 2.2g/cm3
Dispensette™ Organic Bottletop Dispenser
Up to 600mbar
15° to 40°C
1 Year
0.2mL
≤0.5% ±50μL
ISO
Analog
Discharge tube, valve, telescopic filling tube, selection of bottle-top thread adapters, manual, and calibration certificate
59° to 104°F
500mm2/sec.
Accessories