BrandTech™ BRAND™ 1000µL Blue Pipet Tips

Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene

Manufacturer: BrandTech™ 712320

 View more versions of this product

Catalog No. 13-688-645

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Tip Refill
    Material Polypropylene
    Sterility Nonsterile
    Graduations 250, 500, 1000μL
    Tip Style Blue
    Packaging Rack
    Quantity 960 Pack
    Certifications/Compliance DiHEMDA- and oleamide-free
    For Use With (Application) For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
    Volume (Metric) 50 to 1000μL
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit