Learn More
BrandTech™ BRAND™ 200µL Yellow Pipet Tips
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Manufacturer:  BrandTech™ 712315
 View more versions of this product
Catalog No. 13-688-615
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Tip Refill
Nonsterile
20, 100μL
Polypropylene
960 Pack
For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
2 to 200μL
Yellow
Yellow
Rack
DiHEMDA- and oleamide-free