Learn More
BrandTech™ BRAND™ 300µL Pipet Tips
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Manufacturer:  BrandTech™ 702208
 View more versions of this product
Catalog No. 13-688-618
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tip Box
Nonsterile
Polypropylene
96 Pack
For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
5 to 300μL
5-300μL;
Rack
DiHEMDA- and oleamide-free