BrandTech™ BRAND™ 300µL Pipet Tips Available on GSA/VA Contract for Federal Government customers only.
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Manufacturer: BrandTech™ 702353
 View all options for this product
Catalog No. 13-688-620
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Product Type Tip Refill
    Volume (Metric) 5 to 300μL
    Sterility Nonsterile
    Tip Style 5-300μL
    Material Polypropylene
    Packaging Rack
    Quantity 960 Pack
    Certifications/Compliance DiHEMDA- and oleamide-free
    For Use With (Application) For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
    Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
    • Pigment-free
    • Available either in bulk packages or in convenient rack-and-stack formats
  • Description & Specifications
Related Products
Product Content Feedback