BrandTech™ BRAND™ 300µL Pipet Tips
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Manufacturer: BrandTech™ 702533
 View more versions of this product
Catalog No. 13-688-617
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Tip
    Volume (Metric) 5 to 300μL
    Sterility Nonsterile
    Tip Style Precision-molded
    Material Polypropylene
    Packaging Bulk
    Quantity 1000 Pack
    Certifications/Compliance DiHEMDA- and oleamide-free
    For Use With (Application) For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
    Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
    • Pigment-free
    • Available either in bulk packages or in convenient rack-and-stack formats
  • Description & Specifications
Related Products
Product Content Feedback