Learn More
BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 1 to 20uL Available on GSA/VA Contract for Federal Government customers only.
DNA-, RNase-, ATP- and endotoxin-free
Manufacturer:  BrandTech™ 732626
 View more versions of this product
Catalog No. 13-889-166
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Filter Tip
Sterile
1, 20μL
Polypropylene
960 Pack
High translucency, easy ejection and low waste, quick volume check
1 to 20μL
Colorless
Standard
TipRefill, 10 racks of 96 pieces
DNA, RNase, ATP and endotoxin-free