Learn More
BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 50 to 1000uL Available on GSA/VA Contract for Federal Government customers only.
Constructed of high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 50 to 1000µL are sterile and free from biological contaminants. Non self-sealing PE filters allow sample recovery with wetted filters. Thin-walled for high translucency, easy ejection, and low waste. Graduations at 250, 500, and 1000µL provide quick volume check. Available in standard or ultra low retention types.
Manufacturer:  BrandTech™ 732834
 View more versions of this product
Catalog No. 13-889-208
  Check Availability
Add to cart
Specifications
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Non self-sealing PE filters allow sample recovery with wetted filters
  • Thin-walled for high translucency, easy ejection, and low waste.
  • Graduations at 250, 500, and 1000µL for quick volume check.
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format; racked tips are colorless and placed into blue-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties
Description & Specifications
Specifications
Filter Tip
Sterile
250, 500, 1000μL
Ultra Low Retention
TipBox, 10 boxes of 96 pieces
2.75 in.
DNA, RNase, ATP and endotoxin-free
50 to 1000μL
Colorless
Yes
Polypropylene
960 Pack
7cm