Learn More
BrandTech™ BRAND™ BIO-CERT™ nano-cap™ Filter Tips Available on GSA/VA Contract for Federal Government customers only.
DNA-, RNase-, ATP- and endotoxin-free
Manufacturer:  BrandTech™ 732622
 View more versions of this product
Catalog No. 13-889-164
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Filter Tip
Sterile
Standard
TipRefill, 10 racks of 96 pieces
1.45 in.
DNA, RNase, ATP and endotoxin-free
0.1 to 1μL
Colorless
Polypropylene
960 Pack
3.7cm
Sub-micro Transferpette S