BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 0.5 to 20uL Available on GSA/VA Contract for Federal Government customers only.

DNA-, RNase-, ATP- and endotoxin-free

Manufacturer: BrandTech™ 732364

 View more versions of this product

Catalog No. 13-889-218

  Check Availability
Add to Cart

    Specifications

    Sterility Sterile
    Product Type Tip
    Material Polypropylene
    Length (English) 1.81 in.
    Length (Metric) 4.6cm
    Graduations 2, 10μL
    Autoclavable Yes
    Tip Style Ultra Low Retention
    Packaging TipStack, 10 boxes of 96 pieces and two boxes
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    For Use With (Application) High translucency, easy ejection and low waste, quick volume check
    Volume (Metric) 0.5 to 20μL
    Color Colorless

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit