Learn More
BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 1 to 50uL Available on GSA/VA Contract for Federal Government customers only.
DNA, RNase-, ATP- and endotoxin-free
Manufacturer:  BrandTech™ 732326
 View more versions of this product
Catalog No. 13-889-198
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tip
Sterile
Ultra Low Retention
TipBox, 10 boxes of 96 pieces
DNA, RNase, ATP and endotoxin-free
1 to 50μL
Yes
Polypropylene
960 Pack
For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss