BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 1 to 50uL Available on GSA/VA Contract for Federal Government customers only.
DNA, RNase-, ATP- and endotoxin-free
Manufacturer: BrandTech™ 732126
 View more versions of this product
Catalog No. 13-889-160
  Check Availability
Add to Cart
    Specifications
    Product Type Tip
    Material Polypropylene
    Sterility Sterile
    Tip Style Standard
    Packaging TipRefill, 10 racks of 96 pieces
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    Volume (Metric) 1 to 50μL
  • Description & Specifications
Related Products
Product Content Feedback