BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 2 to 200uL Available on GSA/VA Contract for Federal Government customers only.
Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 2 to 200µL are independently certified sterile and free of biological contaminants. Thin-walled for high-translucency, easy ejection, and low waste. Graduations at 20 and 100µL provide quick volume assessment.
Manufacturer: BrandTech™ 732268
 View more versions of this product
Catalog No. 13-889-213
  Check Availability
Add to Cart
    Specifications
    Product Type Tip
    Material Polypropylene
    Sterility Sterile
    Graduations 20, 100μL
    Tip Style Standard
    Packaging TipStack, 10 racks of 96 pieces, two boxes
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    Volume (Metric) 2 to 200μL
    Color Colorless
    • High-purity polypropylene is oleamide- and diHEMDA-free
    • Independently certified sterile and free of biological contaminants
    • Thin-walled for high-translucency, easy ejection and low waste
    • Graduation at 20 and 100µL for quick volume check
    • Available in either standard or ultra low retention types
    • Available in convenient rack-and-stack formats
    • Racked tips are colorless and placed into yellow-colored tip tray
    Ultra low retention tips
    • Produced through special, patented physicochemical process
    • Homogeneous, defect-free surface for low surface tension and low sample retention
    • Reduce sample loss and provide substantially higher reproducibility when working with critical media
    • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
    • High chemical resistance
    • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
  • Description & Specifications
Related Products
Product Content Feedback