Learn More
BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 2 to 200uL Available on GSA/VA Contract for Federal Government customers only.
Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 2 to 200µL are independently certified sterile and free of biological contaminants. Thin-walled for high-translucency, easy ejection, and low waste. Graduations at 20 and 100µL provide quick volume assessment.
Manufacturer:  BrandTech™ 732128
 View more versions of this product
Catalog No. 13-889-161
  Check Availability
Add to cart
Specifications
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • Thin-walled for high-translucency, easy ejection and low waste
  • Graduation at 20 and 100µL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient rack-and-stack formats
  • Racked tips are colorless and placed into yellow-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
Description & Specifications
Specifications
Tip
Sterile
20, 100μL
Polypropylene
960 Pack
2 to 200μL
Colorless
Standard
TipRefill, 10 racks of 96 pieces
DNA, RNase, ATP and endotoxin-free