Learn More
BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 5 to 300uL Available on GSA/VA Contract for Federal Government customers only.
Manufactured from high purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 5 to 300µL are sterile and free of biological contaminants, DNA-, RNase-, ATP- and endotoxin-free. Thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 50, 100 and 300µL provide quick volume assessment. Available in either standard or ultra low retention types.
Manufacturer:  BrandTech™ 732330
 View more versions of this product
Catalog No. 13-889-200
  Check Availability
Add to cart
Specifications
Description
Used for virtually all pipets with yellow color code.
  • High purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • For reverse pipetting, plate washing and working with multichannel pipets
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations marked at 50, 100 and 300µL provide quick volume assessm
  • Available in either standard or ultra low retention types
  • Available in convenient rack-and-stack formats
  • Racked tips are colorless and placed into green-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents or other difficult liquids that can lead to sample loss
  • High chemical resistance.
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties.
Description & Specifications
Specifications
Tip
Sterile
50, 100, 300μL
Ultra Low Retention
TipBox, 10 boxes of 96 pieces
2.08 in.
DNA, RNase, ATP and endotoxin-free
5 to 300μL
Colorless
Yes
Polypropylene
960 Pack
5.3cm
For reverse pipetting, plate washing and working with multichannel pipettes, high translucency, easy ejection and low waste