Learn More
BrandTech™ BRAND™ Dispensette™ III, Digital Adjustable Bottle Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
Uses a floating piston design with no seals to wear or replace, for long service life
Manufacturer:  BrandTech™ 4701350
 View more versions of this product
Catalog No. 13-688-553
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Dispensette™ III Bottletop Dispenser
Up to 2.2g/cm3
Yes
0.1mL
≤0.1% 25μL
59° to 104°F
500mm2/sec.
Digital
≤0.5% ±125μL
Up to 600mbar (vapor pressure)
Discharge tube, 125 to 240mm/170 to 330mm L telescoping filling tube, valve mounting/calibration tool, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual
15° to 40°C
2.5 to 25mL
Adjustable, Easy Calibration, Without SafetyPrime™ valve