Learn More
BrandTech™ BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers
For safe and accurate dispensing of many laboratory liquids, including most acids, bases, salt solutions, scintillation fluids and many organic solvents
Manufacturer:  BrandTech™ 4701121
 View more versions of this product
Catalog No. 13-688-226
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
0.2 to 2mL
Yes
≤0.1% 2μL
Up to 2.2g/cm3
Dispensette™ III Bottletop Dispenser
Up to 600mbar
15° to 40°C
1 Year
0.05mL
≤0.5% ±10μL
GLP, ISO 9000
Analog
SafetyPrime™ valve, 45mm thread, discharge tube, 125 to 240mm L telescoping filling tube, valve mounting/calibration tool, 24, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual
59° to 104°F
500mm2/sec.
Accessories