Learn More
BrandTech™ BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
For safe and accurate dispensing of many laboratory liquids, including most acids, bases, salt solutions, scintillation fluids and many organic solvents
Manufacturer:  BrandTech™ 4701161
 View more versions of this product
Catalog No. 13-688-231
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5 to 50mL
Yes
≤0.1% 50μL
Up to 2.2g/cm3
Dispensette™ III Bottletop Dispenser
Up to 600mbar
15° to 40°C
1 Year
1mL
≤0.5% ±250μL
GLP, ISO 9000
Analog
SafetyPrime™ valve, 45mm thread, discharge tube, 125 to 240mm/170 to 330mm L telescoping filling tube, valve mounting/calibration tool, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual
59° to 104°F
500mm2/sec.
Accessories