BrandTech™ BRAND™ Dispensette™ TA Bottletop Dispensers Available on GSA/VA Contract for Federal Government customers only.
Provides outstanding performance for precise-volume dispensing of high purity chemicals for trace analysis
Manufacturer: BrandTech™ 707955
 View more versions of this product
Catalog No. 13-688-335
  Check Availability
Add to Cart
    Specifications
    Warranty 1 Year
    Product Type Discharge Tube
    For Use With (Equipment) Dispensette™ TA bottletop dispenser
    Includes Integrated valve, platinum iridium valve spring
    Length (English) 3.54 in.
    Length (Metric) 90mm
  • Description & Specifications
Related Products
Product Content Feedback