BrandTech™ BRAND™ Filter Pipet Tips
Suited for applications such as PCR, microbiology and radioisotope work, combine the highest quality filters with premium tips to maximize tip performance by ensuring a great fit and a perfect seal
Manufacturer: BrandTech™ 702120
 View more versions of this product
Catalog No. 13688661
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Sterility Sterile
    Product Type Filter Tip
    Material Polypropylene
    Autoclavable Yes
    Tip Style Filter Tip-Set S
    Packaging 5 filled 96-tip inserts, 1 PP box, 480 tips
    Certifications/Compliance CE IVD 98/79 EC
    Volume (Metric) 0.5 to 10μL
    Color Clear
  • Description & Specifications
Related Products
Product Content Feedback