BrandTech™ BRAND™ Filter Pipet Tips
Suited for applications such as PCR, microbiology and radioisotope work, combine the highest quality filters with premium tips to maximize tip performance by ensuring a great fit and a perfect seal
Manufacturer: BrandTech™ 702164
 View more versions of this product
Catalog No. 13688668
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Filter Tip
    Material Polypropylene
    Packaging 10 boxes of 96 tips, 960 tips
    Sterility Sterile
    Autoclavable Yes
    Tip Style Filter Tip-Box S
    Certifications/Compliance CE IVD 98/79 EC
    Volume (Metric) 5 to 100μL
    Color Clear
  • Description & Specifications
Related Products
Product Content Feedback