BrandTech™ BRANDplates cellGrade™ 1536-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: BrandTech™ 782000

 View more versions of this product

Catalog No. 14-386-009

  Check Availability
Add to Cart

    Specifications

    Lid Yes,Yes
    For Use With (Application) Cultivation of adherent cell lines
    Format 1536-well,1536-well
    No. of Wells 1536
    Sterile Yes
    Surface Coating Carboxyl and Hydroxyl Groups
    Type Premium 1536-Well Microplate,Premium 1536-Well Microplate
    Certifications/Compliance ANSI, SLAS
    Well Design Flat (F)
    Closure Yes
    Color Transparent
    Material Polystyrene,Polystyrene
    Volume (Metric) 10μL,10μL
    Wells 1536

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit