Learn More
BrandTech™ BRANDplates cellGrade™ 1536-Well Microplates
BRANDplates™ for cell culture are available in a variety of surface treatments for use in the culture of a wide range of cell types.
Manufacturer:  BrandTech™ 782002
 View more versions of this product
Catalog No. 14-386-011
Add to cart
Description
Description
  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free; noncytotoxic
  • Endotoxins are <0.01 EU/mL
  • For standard cell culture applications
  • Individually wrapped with lid
Specifications
Specifications
Flat
Black
Polystyrene
Carboxyl and Hydroxyl Groups
10μL
Sterile
ANSI, SLAS
Cultivation of adherent cell lines
1536
With Lid
Premium 1536-Well Microplate