Learn More
BrandTech™ BRANDplates cellGrade™ 384-well Cell Culture Microplates
Standard plate for the cultivation of adherent cell lines with hydrophillic surface.
Manufacturer:  BrandTech™ 781987
 View more versions of this product
Catalog No. 14-386-051
Add to cart
Description
Description
  • Polystyrene construction
  • Sterile
  • Lids included
  • Standard plate for the cultivation of adherent cell lines
  • PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible
  • Surface is hydrophilic compared with non-treated PS
  • Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells
  • Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Specifications
F Bottom
Black
Cultivation of adherent cell lines
PS (Polystyrene)
Yes
With Lid
120μL
Sterile
ANSI, SLAS
Yes
384 well
384
Carboxyl and Hydroxyl Groups
50/PK
Transparent bottom