Learn More
BrandTech™ BRANDplates cellGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Designed for the cultivation of adherent cell lines
Manufacturer:  BrandTech™ 781961
 View more versions of this product
Catalog No. 13-882-245
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Yes
Cultivation of adherent cell lines
Polystyrene
Yes
96-Well Microplate
Conical (V)
ISO 11137, DIN EN ISO 10993
Transparent
96-well
96
Carboxyl and Hydroxyl Groups
360μL