Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
1/Pk., 50/Cs.
Description
- Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
- Easily binds serum components to for indirect adhesion of cells
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- DNA-, DNase-, RNase-free
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
Certifications/Compliance | ISO 11137, DIN EN ISO 10993 |
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Binding Type | Carboxyl and Hydroxyl Groups |
Color | Black, Clear Bottom |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Sterile |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction