BrandTech™ BRANDplates cellGrade™ plus 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Designed for the cultivation of cells in reduced-serum media and fastidious cell lines

Manufacturer: BrandTech™ 782031

 View more versions of this product

Catalog No. 13-882-256

  Check Availability
Add to Cart

    Specifications

    Lid Yes,Yes
    For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
    Format 96-well,96-well
    Material Polystyrene,Polystyrene
    No. of Wells 96
    Sterile Yes
    Surface Coating Hydrophilic
    Type Plus 96-Well Microplate,Plus 96-Well Microplate
    Volume (Metric) 330μL,330μL
    Certifications/Compliance ISO 11137, DIN EN ISO 10993
    Closure Yes
    Color Black
    Wells 96

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit