BrandTech™ BRANDplates cellGrade™ premium 96-Well Microplates

Plates with surface characteristics for the most demanding cell lines

Manufacturer: BrandTech™ 782085

 View more versions of this product

Catalog No. 13-882-258

  Check Availability
Add to Cart

    Specifications

    Lid Yes,Yes
    For Use With (Application) Cultivation of cell lines with the highest demands on their environment
    Format 96-well,96-well
    Material Polystyrene,Polystyrene
    No. of Wells 96
    Sterile Yes
    Surface Coating Poly-D-Lysine Equivalent
    Type Premium 96-Well Microplate,Premium 96-Well Microplate
    Volume (Metric) 350μL,350μL
    Certifications/Compliance ISO 11137, USP 29
    Closure Yes
    Color White
    Wells 96

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit