Learn More
BrandTech™ BRANDplates cellGrade™ premium 96-Well Microplates
Plates with surface characteristics for the most demanding cell lines
Manufacturer:  BrandTech™ 782088
 View more versions of this product
Catalog No. 13-882-259
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Yes
Cultivation of cell lines with the highest demands on their environment
Polystyrene
Yes
Premium 96-Well Microplate
ISO 11137, USP 29
Black
96-well
96
Poly-D-Lysine Equivalent
350μL