Learn More
BrandTech™ BRANDplates hydroGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution
Manufacturer:  BrandTech™ 781783
 View more versions of this product
Catalog No. 13-882-229
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
No
No
Immobilization of hydrophilic molecules
Polystyrene
No
96-Well Microplate
Round
ANSI, SLAS
Transparent
96-well
96
Hydrophilic
350μL